Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Gb3
- Antigen symbol:Gb3
- Clonality:Monoclonal
- Clone:BGR23
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Size:100 μg
- Format:Ascites Fluid
- Storage temperature:Store at -20 ºC to -80 ºC (Preferably at -80 ºC). It is not recommended storing at 4 ºC. Avoid repeated freezing and thawing.
- Shipping temperature:-80°C
- Immunogen:Purified glycolipid adsorbed to Salmonella Minnesota:Gb3
- Cat. no.:TCA2822-VIAL
- Packaging:Vial
- Supplier No.:A2822-VIAL
Biotinylated mouse monoclonal antibody/IgG2b, κ-chain
Type: Primary
Antigen: Gb3
Clonality: Monoclonal
Clone: BGR23
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: