Order Entry
Canada
ContactUsLinkComponent
SensoLyte® 490 HCV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® 490 HCV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-146
Supplier:  Anaspec
CAS Number:  
SensoLyte® 490 HCV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-146
Supplier:  Anaspec
Supplier Number:  AS-71126
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    SensoLyte® 490 HCV Protease Assay Kit, Fluorimetric
  • Size:
    500 Assays
  • Cat. no.:
    103010-146
  • Storage Temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-71126

Specifications

About this item

HCV protease is identified as an important drug-screening target.

The SensoLyte® 490 HCV Protease Assay Kit uses an optimized FRET peptide substrate for the continuous measurement of HCV NS3/4a activity. This FRET substrate is cleaved specifically by HCV NS3/4a protease thereby liberating the C-terminal peptide-fluorophore fragment from the proximity quenching effect of the dark quencher, resulting in a more than 10-fold increase in fluorescence. This assay can detect HCV NS3/4a protease activity at 1 pmole.