Order Entry
Canada
ContactUsLinkComponent
SensoLyte® Green SIRT1 Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® Green SIRT1 Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-516
Supplier:  Anaspec
CAS Number:  
SensoLyte® Green SIRT1 Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-516
Supplier:  Anaspec
Supplier Number:  AS-72156
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. no.:
    103010-516
  • Storage Temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72156

Specifications

About this item

Histone deacetylase (HDAC) enzymes act as transcriptional repressors of genes through the deacetylation of lysine residues on histone proteins

The SensoLyte® Green SIRT1 Assay Kit provides a convenient, two-step homogeneous procedure for measuring sirtuin 1 activity using a fluorogenic substrate. In the first step, an acetylated substrate is incubated with SIRT1 containing samples. Deacetylation of substrate first sensitizes it, so that, in the second step, mixing with the SIRT developer generates a fluorophore, which can then be detected at Ex/Em= 490 nm/520 nm. This kit can be used for high throughput screening of SIRT1 inhibitors/activators. The long wavelength fluorescence of SIRT1 substrate is less interfered by the autofluorescence of cell components and test compounds. Assays are performed in a standard 96-well format.

Sirtuins (SIRTs) are a unique class of deacetylases (class III HDACs) that require NAD as a cofactor. Sirtuin 1 (SIRT1) is involved in regulation of tumor suppressor p53 and inhibition of apoptosis. It also represents a target for treatment of age-related diseases and type II diabetes.