If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 Assays
- Cat. no.:103010-342
- Storage Temperature:– 78 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-72040
Specifications
About this item
Renin, a highly specific aspartyl protease, cleaves angiotensinogen, to yield angiotensin I, which is further converted into angiotensin II by ACE (Angiotensin Converting Enzyme)
The SensoLyte® 520 Renin Assay Kit provides a convenient assay for high throughput screening of renin inhibitors and for continuous assay of renin activity using a 5-FAM/QXL®520 fluorescence resonance energy transfer (FRET) peptide. In the FRET peptide the fluorescence of 5-FAM is quenched by QXL®520. Upon cleavage into two separate fragments by renin, the fluorescence of 5-FAM is recovered, and can be monitored at excitation/emission = 490/520 nm. This assay is about fifty fold more sensitive than an EDANS/DABCYL-based assay and can detect 0.8 ng/mL renin.
Angiotensin II constricts blood vessels leading to increased blood pressure. It also increases the secretion of ADH and aldosterone, and stimulates the hypothalamus to activate the thirst reflex. Since an overactive renin-angiotensin system leads to hypertension, renin is an attractive target for the treatment of this disease.