If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:500 Assays
- Cat. no.:103010-144
- Storage Temperature:– 20 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-71124
Specifications
About this item
The SensoLyte® Green Protease Assay Kit is widely used for detection of generic protease activities. The kit uses a casein derivative that is heavily labeled with a rhodamine derivative, resulting in almost total quenching of the conjugate's fluorescence. Protease-catalyzed hydrolysis relieves this quenching conjugate, yielding brightly green fluorescent dye-labeled peptides. The increase in fluorescence intensity is directly proportional to protease activity (Ex/Em=488/520 nm). The SensoLyte® protease assay kits do not require any separation steps and can be used to continuously measure the kinetics of a variety of exopeptidases and endopeptidases.