Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:96 Assays
- Cat. no.:103001-340
- Storage Temperature:+4 °C
- Includes:One 96-well plate, essential components and detailed protocol and references
- Supplier No.:AS-55523
SensoLyte® Anti-PLP (178-191) IgG Quantitative ELISA Kit (Mouse) is optimized to detect mouse anti-PLP (178-191) IgG. This kit is useful to researchers who wish to determine the amount of anti-PLP (178-191) antibody present in biological samples, and can help provide information on the role it plays in the development and treatment of EAE, an animal model for MS pathogenesis. Wells are pre-coated with PLP (178-191) peptide and pre-blocked with BSA. The amount of anti-PLP (178-191) IgG in mouse serum or cerebrospinal fluid is quantified using ELISA (Abs=450 nm). Ample materials and reagents are provided to perform 96 assays.