If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:100 Assays
- Cat. no.:103010-456
- Storage Temperature:– 78 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-72124
Plasmin belongs to the family of serine proteases
The SensoLyte® AFC Plasmin Activity Assay Kit provides a convenient assay for high throughput screening of plasmin inhibitors and inducers or for continuous assay of plasmin activity. The assay utilizes a fluorogenic peptide, which upon enzyme cleavage releases the AFC (7-amido-4-trifluoromethylcoumarin) fluorophore. The fluorescent signal can be measured at Ex/ Em =380 nm/500 nm. This assay can detect picogram level of plasmin activity. Assays are performed in a convenient 96-well microplate format.
It plays a key role in fibrinolysis by dissolving fibrin in blood clots. Besides fibrinolysis, plasmin is also involved in such physiological and pathological processes as wound healing, liver repair, and the maintenance of liver homeostasis.