ContactUsLinkComponent
SensoLyte® Green Pin1 Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® Green Pin1 Assay Kit (Fluorimetric), AnaSpec Inc.
  103008-974
 :  Anaspec
 :  
SensoLyte® Green Pin1 Assay Kit (Fluorimetric), AnaSpec Inc.
  103008-974
 :  Anaspec
 :  AS-72240
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    100 Assays
  • Cat. no.:
    103008-974
  • Storage Temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72240
Peptidyl-prolyl isomerase (PPIase) Pin1 catalyzes cis/trans isomerization of the phospho-Serine/Threonine-Proline peptide bonds
The SensoLyte® Green Pin1 Assay Kit uses a fluorogenic substrate in the cis isoform. Pin1 changes substrate into trans conformation, which is then readily cleaved to generate fluorescent signal which can be monitored at Ex/Em=490/520nm. Increase in fluorescence intensity is directly proportional to the Pin1 activity.
Pin1 consists of short N-terminal protein-protein interaction domain that allows enzyme to bind phosphoproteins and longer C-terminal izomerase domain. Pin1 has many biological substrates, plays critical role in cell-cycle regulation, and up-regulated in many human cancers. Recently, Pin1 was linked to the Alzheimer’s disease pathogenesis. Pin1 can bind phosphorylated Thr-212/231 residues of Tau protein and increase its deposphorylation that may prevent formation of Tau neurofibrilary tangles.