Order Entry
Canada
ContactUsLinkComponent
SensoLyte® MG Phosphate Assay Kit (Colorimetric), AnaSpec
SensoLyte® MG Phosphate Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-114
Supplier:  Anaspec
CAS Number:  
SensoLyte® MG Phosphate Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-114
Supplier:  Anaspec
Supplier Number:  AS-71103
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1000 Assays
  • Cat. no.:
    103010-114
  • Storage Temperature:
    RT
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-71103

Specifications

About this item

The MG (Malachite Green) Phosphate Assay Kit is based on quantification of the green complex formed between Malachite Green, molybdate, and free orthophosphate

The kit has been used for assaying phosphatase, phospholipase or lipase, nucleoside triphosphatase (ATPase, GTPase, etc.) and phytase. The kit has the following components: Highly purified Malachite Green reagent, Protein phosphatase inhibitor, Assay buffer, A 'mix and read' assay protocol that is compatible with HTS liquid handling instruments with Z' factors of 0.7 to 0.9.

The rapid color formation from the reaction can be conveniently measured on a spectrophotometer (600 to 660 nm) or on a plate reader. The non-radioactive colorimetric assay kits have been optimized to offer superior sensitivity and prolonged shelf life. The assay is simple and fast, involving a single addition step for phosphate determination. Assays can be performed in tubes, cuvettes, or multi-well plates. The assays can be conveniently performed in 96 or 384-well plates for high-throughput screening of enzyme inhibitors.