Order Entry
Canada
ContactUsLinkComponent
SensoLyte® Anti-Rat MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
SensoLyte® Anti-Rat MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
Catalog # 102998-094
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® Anti-Rat MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
Catalog # 102998-094
Supplier:  Anaspec Inc
Supplier Number:  AS-55157
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    96 Assays
  • Cat. no.:
    102998-094
  • Storage Temperature:
    +4 °C
  • Includes:
    One 96-well plate, essential components and detailed protocol and references
  • Supplier No.:
    AS-55157

Specifications

About this item

This kit is optimized to detect anti-rat MOG (1-125) IgG. Wells are pre-coated with recombinant rat MOG (1-125) protein and pre-blocked with BSA. The amount of anti-rat MOG IgG in serum or cerebrospinal fluid is quantified using ELISA. Ample materials and reagents are provided to perform 96 assays.