Order Entry
Canada
ContactUsLinkComponent
SensoLyte® Anti-Mouse MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
SensoLyte® Anti-Mouse MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
Catalog # 102998-092
Supplier:  Anaspec
CAS Number:  
SensoLyte® Anti-Mouse MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
Catalog # 102998-092
Supplier:  Anaspec
Supplier Number:  AS-55156
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    96 Assays
  • Cat. no.:
    102998-092
  • Storage Temperature:
    +4 °C
  • Includes:
    One 96-well plate, essential components and detailed protocol and references
  • Supplier No.:
    AS-55156

Specifications

About this item

This kit is optimized to detect anti-mouse MOG (1-125) IgG. Wells are pre-coated with recombinant mouse MOG (1-125) protein and pre-blocked with BSA. The amount of anti-mouse MOG IgG in serum or cerebrospinal fluid is quantified using ELISA. Ample materials and reagents are provided to perform 96 assays.