Order Entry
Canada
ContactUsLinkComponent
SensoLyte® Rh110 Matriptase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® Rh110 Matriptase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103008-976
Supplier:  Anaspec
CAS Number:  
SensoLyte® Rh110 Matriptase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103008-976
Supplier:  Anaspec
Supplier Number:  AS-72241
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. no.:
    103008-976
  • Storage Temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72241

Specifications

About this item

Matriptase (also known as MT-SP1, ST14, TADG-15 and epithin) is a trypsin-like protease from the family of type II transmembrane serine proteases

The SensoLyte® Rh110 Matriptase Assay Kit provides a convenient assay for screening of enzyme inhibitors and activators or for continuous assay of enzyme activity using a fluorogenic substrate. Upon matriptase cleavage, this substrate generates the Rh110 (Rhodamine 110) fluorophore which has a bright green fluorescence and can be detected at excitation/emission=496 nm/520 nm. The longer wavelength spectra and higher extinction coefficient of Rh110 provide greater sensitivity and less interference from other reaction components. This assay can detect as low as 0.15 ng/mL active Matriptase.

The spectrum of known Matriptase substrates includes extracellular matrix proteins, cell adhesion molecules, ion channels, growth-factor-like proteins and other proteases. Its actions can result in protein processing, activation or degradation. Matriptase is widely expressed in virtually all epithelium and is specifically found in tumors of epithelial origin. Matriptase has been implicated in carcinogenesis including ovarian, prostate and cervical cancers and is considered a drug target for developing cancer therapeutic agent.