If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 Assays
- Cat. no.:103010-518
- Storage Temperature:– 78 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-72157
Specifications
About this item
GST (Glutathione S-Transferase) enzymes, a family of isozymes plays a crucial role in body defense mechanisms against toxins and carcinogens
The SensoLyte® GST Activity Assay Kit utilizes an optimized method for detection of enzyme activity in biological samples. The kit provides substrate for GST enzyme, which upon reaction with reduced glutathione yields a fluorescent glutathione adduct. This produces bright blue fluorescence that can be monitored at excitation/emission =390 nm/480 nm.
GSTs carry out their functions mostly by mediating their conjugation with reduced glutathione on substrates such as pharmaceutical and environmental pollutants.