Order Entry
Canada
ContactUsLinkComponent
 
SensoLyte® ONPG β-Galactosidase Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-474
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® ONPG β-Galactosidase Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-474
Supplier:  Anaspec Inc
Supplier Number:  AS-72134
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. no.:
    103010-474
  • Storage Temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72134

Specifications

About this item

β-galactosidase, encoded by the lacZ gene in E. coli, is widely used as a reporter enzyme to study gene expression, protein-protein interaction and normalization of transfection efficiency in mammalian cells.

The SensoLyte® ONPG β-Galactosidase Assay Kit utilizes the widely used chromogenic substrate, o-nitrophenyl β-D-galactopyranoside (ONPG) to determine the activity of β-galactosidase in transfected cells or tissues. In the presence of β-galactosidase, the colorless compound is hydrolyzed into o-nitrophenol, producing a yellow color that can be detected at absorbance 420 nm. The intensity of the color produced is proportional to β-galactosidase activity.