Order Entry
Canada
ContactUsLinkComponent
SensoLyte® FDG β-Galactosidase Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® FDG β-Galactosidase Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-472
Supplier:  Anaspec
CAS Number:  
SensoLyte® FDG β-Galactosidase Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-472
Supplier:  Anaspec
Supplier Number:  AS-72133
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. no.:
    103010-472
  • Storage Temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72133

Specifications

About this item

β-galactosidase, encoded by the lacZ gene in E

The SensoLyte® FDG β-Galactosidase Assay Kit provides a sensitive fluorogenic substrate for detecting β-galactosidase activity in tranfected cells. In the presence of β-galactosidase, the colorless fluorescein di-β-D galactopyranoside (FDG) substrate is hydrolyzed into a fluorescein that can be detected at Ex/Em = 490/520 nm.

coli, is widely used as a reporter enzyme to study gene expression, protein-protein interaction and normalization of transfection efficiency in mammalian cells.