If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 Assays
- Cat. no.:103008-962
- Storage Temperature:– 20 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-72085
Specifications
About this item
TACE (tumor necrosis factor-alpha converting enzyme), α-secretase, or ADAM17 is a member of the ADAM family of proteases that contain both disintegrin and metalloprotease (catalytic) domains
SensoLyte® 520 TACE Activity Assay Kit uses a 5-FAM (fluorophore) and QXL® 520 (quencher) labeled FRET peptide substrate for continuous measurement of enzyme activity. In the intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage of the FRET peptide by the active enzyme, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, 5-FAM/QXL® 520 based FRET peptide has less interference from the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The kit can be used to detect the activity of TACE or screen for TACE inhibitors. The assays are performed in a convenient 96-well format.
This enzyme is involved in sperm-egg binding and fusion, muscle cell fusion, neurogenesis, modulation of Notch receptor and ligand processing, and processing of the pro-inflammatory cytokine, TNF α. TACE is currently being explored as a target for anti-inflammatory drugs.