If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:500 Assays
- Cat. no.:103010-162
- Storage Temperature:–20 °C
- Includes:The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-71135
Specifications
About this item
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 490 MMP-13 Assay Kit is a complete assay system designed to continuously analyze MMP-13 activities or to screen MMP-13 inhibitors by using a fluorogenic MMP-13 substrate. The assays are performed in a convenient 96-well or 384-well microplate format. The kit provides speed, convenience and accuracy in analyzing MMP-13 activity and screening MMP-13 inhibitors. Members of the MMP family have poor substrate sequence specificity, making it difficult to use a peptide substrate alone to differentiate the activity of a particular MMP from other MMPs. If several MMPs are coexisting in your samples and you would like to specifically measure MMP-13 activity, please choose the SensoLyte® Plus MMP-13 Assay Kit.
MMP-13 (collagenase-3, rat collagenase) is involved in quite a few diseases such as arthritis and asthma.