Order Entry
Canada
ContactUsLinkComponent
SensoLyte® 490 MMP-9 Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 490 MMP-9 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-160
Supplier:  Anaspec
CAS Number:  
SensoLyte® 490 MMP-9 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-160
Supplier:  Anaspec
Supplier Number:  AS-71134
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. no.:
    103010-160
  • Storage Temperature:
    –20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-71134

Specifications

About this item

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components

The SensoLyte® 490 MMP-9 Assay Kit is a complete assay system designed to continuously analyze MMP activity or to screen for MMP-9 inhibitors using a fluorogenic MMP-9 substrate. The assays are performed in a convenient 96-well or 384-well microplate format. The kit provides speed, convenience and accuracy in MMP-9 analysis. Members of the MMP family have poor substrate sequence specificity, making it difficult to use a peptide substrate alone to differentiate the activity of a particular MMP from other MMPs. If several MMPs are coexisting in your samples and you would like to specifically measure MMP-9 activity, please choose the SensoLyte® Plus MMP-9 Assay Kit.

MMP-9 (gelatinase-B, collagenase-IV) is involved in quite a few diseases such as a cancer, angiogenesis, alopecia and metastasis.