Order Entry
Canada
ContactUsLinkComponent
SensoLyte® Plus 520 MMP-9 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
SensoLyte® Plus 520 MMP-9 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
Catalog # 103010-290
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® Plus 520 MMP-9 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
Catalog # 103010-290
Supplier:  Anaspec Inc
Supplier Number:  AS-72017
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    96 Assays
  • Cat. no.:
    103010-290
  • Storage Temperature:
    –20 °C
  • Includes:
    Kit includes a 96-well plate, essential components and detailed protocol and references
  • Supplier No.:
    AS-72017

Specifications

About this item

The SensoLyte® Plus 520 MMP-9 Assay Kit is designed for specifically detecting MMP-9 activity in biological samples which may contain multiple MMPs, such as culture medium, serum, plasma, synovial fluid, and tissue homogenate. A specific anti-MMP-9 monoclonal antibody is used in combination with a MMP fluorogenic substrate, 5-FAM/QXL®520 FRET peptide. The fluorescence signal is monitored at Ex/Em=490 nm/520 nm upon MMP-9-induced cleavage of the FRET substrate. Ample materials are provided to perform 96 assays in a 96-well format.