If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:16 x 20 Assays
- Cat. no.:103010-248
- Storage Temperature:–20 °C
- Includes:16 different MMP substrates, essential components and detailed protocol and references
- Supplier No.:AS-71170
Specifications
About this item
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
SensoLyte® 520 MMP Substrate Sampler Kit contains a series of FRET peptide substrates for use as fluorogenic indicators to assay MMP protease activities. It provides the best solution for identifying a suitable fluorogenic substrate for a particular MMP assay. All the MMP substrates in the kit are also available in bulk packages.
They are involved in diseases such as cancer, arthritis, asthma, angiogenesis, alopecia and metastasis. They are also identified as drug-screening targets.