Freeze Substitution Kit, Electron Microscopy Sciences
About this item
For dehydration and fixation of rapidly frozen cells.
- Excellent freeze substitution results in as little as 90 minutes
- Extend the time for larger samples to 3 hours by putting the lid on
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete.
With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
2 Options Available Below
Product Details & Documents
For dehydration and fixation of rapidly frozen cells.
- Excellent freeze substitution results in as little as 90 minutes
- Extend the time for larger samples to 3 hours by putting the lid on
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete.
With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
Specifications for Product Family
Specifications
- Catalog No:
- 103306-432
- 103307-138
- Model:
- 34500-S
- 34500
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- 103306-432Freeze substitution kit without shakerFreeze substitution kit without shakerEach$3,404.82Kit of 1$3,404.82Supplier Number: 34500-SSpecifications:
More
Specifications
Cat. No.103306-432Model34500-S - 103307-138Freeze substitution kit with shakerSupplier Number: 34500Specifications:
More
Specifications
Cat. No.103307-138Model34500
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)