These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
The anti-magnetic steel ergonomic tweezers have a soft electro static discharge (ESD) grip. The red rubber handle has very high resistivity and a comfortable grip.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Length is 109 mm.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.