Order Entry
Canada
ContactUsLinkComponent
Glycerol Mounting Medium, Electron Microscopy Sciences
Glycerol Mounting Medium, Electron Microscopy Sciences
  103305-556
 :  
Glycerol Mounting Medium, Electron Microscopy Sciences
  103305-556
 :  17989-41
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Glycerol Mounting Medium
  • Size:
    100 ml
  • Cat. no.:
    103305-556
  • Supplier No.:
    17989-41

 

 

Aqueous mounting medium with glycerol for preserving fluorescence of tissue and cell smears.

  • Useful for immunofluorescence
  • Does not contain phenylenediamine
  • Refractive Index: 1.472

This media is excellent for preserving thick sections that can not be mounted with mounting medium or fresh frozen tissues that contain a lot of lipids, e.g. brain tissue. It is also useful for preserving small embryos. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy dyes, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during prolonged storage at 4°C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes. It is used for mounting tissues and cell for IHC and for Immunofluorescence (IF) Reagent: Ready to use mounting medium for IHC and immunofluorescence (IF)
Storage: 2-8°C is recommended, Protect from light.

 : Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC).