Order Entry
Canada
ContactUsLinkComponent
AnaTag™ R-PE Labeling Kit, AnaSpec Inc.
AnaTag™ R-PE Labeling Kit, AnaSpec Inc.
Catalogue # 103010-184
Fournisseur:  Anaspec
Numéro CAS:  
AnaTag™ R-PE Labeling Kit, AnaSpec Inc.
Catalogue # 103010-184
Fournisseur:  Anaspec
Supplier Number:  AS-72113
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Size:
    1 Labeling Reaction
  • Cat. no.:
    103010-184
  • Storage Temperature:
    +4 °C
  • Includes:
    This kit is optimized for conjugating R-phycoerythrin (R-PE) to antibody providing ample materials to perform one conjugation reaction for labeling up to 1 mg antibody
  • Supplier No.:
    AS-72113

Spécifications

À propos de cet article

The AnaTag™ R-PE Labeling Kit is optimized for use in the conjugation of R-Phycoerythrin (R-PE) to antibodies. R-PE, a fluorescent protein from phycobiliprotein family, has broad absorption bands with peaks at 565 nm (eM =1.96 x 106 M-1cm-1), 498 (eM =1.53 x 106 M-1cm-1), and 539 nm (eM =1.62 x 106 M-1cm-1); it therefore can be excited with versatile excitation sources. Its maximal emission can be detected at 578 nm. The broad excitation spectrum also provides the advantage of multi-color immunofluorescent staining or cell sorting. R-PE and the closely related B-PE are the most intensely fluorescent phycobiliproteins having orange fluorescence. They are significantly brighter and more photostable than conventional organic fluorophores. The AnaTagTM R-PE Labeling Kit contains SH-reactive R-PE. SMCC modified R-PE reacts with the thiol groups of target antibody without the need for additional activation, thus simplifying the conjugation protocol. The amount of R-PE supplied in this kit is sufficient for labeling up to 1 mg of antibody. The kit provides all reagents, purification columns and a detailed step-by-step protocol. Native and SMCC-activated R-PE are also available as separate products.