Orders LinkContactUsLinkComponent
Forensic Field Sampler, Electron Microscopy Sciences
Forensic Field Sampler, Electron Microscopy Sciences
Forensic Field Sampler, Electron Microscopy Sciences
Certificates
Forensic Field Sampler, Electron Microscopy Sciences
Forensic Field Sampler, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Collect forensic evidence with minimum interference and/or contamination from the sampler.
  • Gunshot residue (GSR)
  • Powder and fiber samples
  • Paint chips
  • Particle sampling
  • Glass fragments
Glass specimen vial (25 OD x 55 mm high) with a high-purity certified 12.7 mm SEM pin stub specimen mount, both with and without an adhesive carbon conductive tab, mounted into the plastic cap
Choice of standard 12.7 mm SEM pin mount with 8 mm pin or 12.7 mm SEM pin mount with shorter 6 mm pin (ZEISS/LEO). Ideal for SEM specimen preparation on the most frequently used SEM pin stubs, facilitating easy carbon coating for SEM/EDX investigation. The Forensic Field Sampler can be securely stored in the glass specimen vial and easily shipped with the cap holding the sampler stub in place. Adhesive carbon tabs secure the evidence material.
This Forensic Field Sampler has been designed to collect forensic evidence with minimum interference and/or contamination from the sampler. Three versions are available: without adhesive carbon tab, with ready-to-use adhesive carbon tab, or with covered carbon tab (plastic cover needs to be removed prior to use).
Product Family Options
Product Information
  • Pin size
    Availability
    Price
  • 103306-164
    Pin Stub without Adhesive Carbon Tab
    Pin Stub without Adhesive Carbon Tab
    6 mm
    Log in for availability
    Log in for availability
    Each
    Each - $497.40
    $497.40
    Each
       $497.40
    Supplier Number: 76480-34
  • 103306-184
    Pin Stub with Adhesive Carbon Tab
    Pin Stub with Adhesive Carbon Tab
    6 mm
    Log in for availability
    Log in for availability
    Each
    Each - $648.01
    $648.01
    Each
       $648.01
    Supplier Number: 76480-35
  • 103306-186
    Pin Stub with Adhesive Carbon Tab, Clear Plastic Cover
    Pin Stub with Adhesive Carbon Tab, Clear Plastic Cover
    6 mm
    Log in for availability
    Log in for availability
    Each
    Each - $648.01
    $648.01
    Each
       $648.01
    Supplier Number: 76480-36
  • 103306-162
    Pin Stub without Adhesive Carbon Tab
    Pin Stub without Adhesive Carbon Tab
    8 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $453.29
    $453.29
    Log in for availability
    Kit of 100
    Kit of 100 - $453.29
    $453.29
    Each
       $453.29
    Kit of 100
       $453.29
    Supplier Number: 76480-31
  • 103306-168
    Pin Stub with Adhesive Carbon Tab
    Pin Stub with Adhesive Carbon Tab
    8 mm
    Log in for availability
    Log in for availability
    Each
    Each - $603.66
    $603.66
    Each
       $603.66
    Supplier Number: 76480-32
  • 103306-170
    Pin Stub with Adhesive Carbon Tab, Clear Plastic Cover
    Pin Stub with Adhesive Carbon Tab, Clear Plastic Cover
    8 mm
    Log in for availability
    Log in for availability
    Each
    Each - $603.66
    $603.66
    Each
       $603.66
    Supplier Number: 76480-33
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)