Mouse IgG2b Isotype Control [clone: MPC-11]
100 μg
Catalog # 75843-388
Supplier: Biogems
Supplier #: 43922-25-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The MPC-11 monoclonal antibody is expressed by a cell line adapted from the Merwin Plasmacytoma-11, and has unknown specificity. The MPC-11 line was obtained by intraperitoneal implantation into a BALB/c Mouseof a Millipore diffusion chamber and is used as an Isotype Control for Mouse IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MPC-11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Specifications
- Size:100 μg
- Cat. No.:75843-388
- Clonality:Monoclonal
- Flow cytometry:Yes
- Reactivity:Mouse
- Format:SAFIRE Purified
- Antibody type:Control
- Form:liquid
- Isotype:IgG2b
- Host:Mouse
- Antigen name:IgG2b Isotype Control (MPC-11)
- Clone:MPC-11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



