Mouse IgG2a Isotype Control [clone: C1.18.4]
75842-484
- Antibody type:Control
- Antigen name:IgG2a Isotype Control
- Clonality:Monoclonal
- Clone:C1.18.4
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Cat. no.:75842-484
- Supplier No.:43912-25-100
The C1.18.4 monoclonal antibody is used as an Isotype Control for MouseIgG2a antibodies. It has unknown specificity and occurred spontaneously in a C3H mouse.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: C1.18.4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse