Mouse IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: C1.18.4]
Catalog # 75842-488
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:C1.18.4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. no.:75842-488
- Supplier No.:43912-50-100
Specifications
About this item
The C1.18.4 monoclonal antibody is used as an Isotype Control for MouseIgG2a antibodies. It has unknown specificity and occurred spontaneously in a C3H mouse.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse