To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. no.:75843-390
- Supplier No.:43922-40-100
The MPC-11 monoclonal antibody is expressed by a cell line adapted from the Merwin Plasmacytoma-11, and has unknown specificity. The MPC-11 line was obtained by intraperitoneal implantation into a BALB/c Mouseof a Millipore diffusion chamber and is used as an Isotype Control for MouseIgG2b antibodies.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MPC-11
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse