Rat IgG1 Isotype Control (BG Violet 450) [clone: HRPN]
Catalog # 75842-470
Supplier: Biogems
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. no.:75842-470
- Supplier No.:42813-40-100
Specifications
About this item
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat