Order Entry
Canada
ContactUsLinkComponent
Rat IgG2b Isotype Control (BG Violet 450) [clone: LTF-2]
Rat IgG2b Isotype Control (BG Violet 450) [clone: LTF-2]
Catalog # 75842-450
Supplier:  Biogems
Rat IgG2b Isotype Control (BG Violet 450) [clone: LTF-2]
Catalog # 75842-450
Supplier:  Biogems
Supplier Number:  41823-40-100

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    LTF-2
  • Conjugation:
    BG Violet 450
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Rat
  • Size:
    100 μg
  • Form:
    liquid
  • Cat. no.:
    75842-450
  • Supplier No.:
    41823-40-100

Specifications

About this item

The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rat