Rat IgG2a, kappa Isotype Control (BG Violet 450) [clone: 2A3]
Catalog # 75842-428
Supplier: Biogems
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. no.:75842-428
- Supplier No.:41813-40-100
Specifications
About this item
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Rat