Mouse IgG1, kappa Isotype Control [clone: MOPC-21]
Catalog # 75841-340
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:100ug
- Form:liquid
- Cat. no.:75841-340
- Supplier No.:44212-20-100
Specifications
About this item
MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse