Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 61D3]
75841-752
- Antibody type:Primary
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:61D3
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen synonyms:Mo2|LPS-R
- Cat. no.:75841-752
- Supplier No.:06211-60-25
The 61D3 monoclonal antibody specifically reacts with CD14, a cell surface anchored glycoprotein that is primarily expressed on monocytes, inerfollicular macrophages, and a subset of dendritic cells. CD14 associates with MD-2 (LY-96) and TLR4 to form a receptor complex, which signals specifically in response to bacterial lipopolysaccharide (LPS) binding. The CD14/MD-2/TLR4 receptor complex signals via MyD88, TIRAP, and TRAF6, and ultimately activates NF-kappaB.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human