Orders LinkContactUsLinkComponent
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
Certificates
JEOL SEM Mount, Electron Microscopy Sciences
JEOL SEM Mount, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
We offer a complete line of SEM specimen mounts.
  • For scanning electron microscopy
  • Standard lathe finish and economically priced
  • Made from ultra-pure aluminum
Head: 50 mm diameter
Product Family Options
Product Information
  • Size
    Material
    Availability
    Price
  • 103301-980
    SEM Mount JEOL
    SEM Mount JEOL
    50 x 5 mm
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $144.77
    $144.77
    Each
       $144.77
    Supplier Number: 75705
  • 103306-174
    SEM Mount JEOL
    SEM Mount JEOL
    50 x 5 mm
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $1,281.28
    $1,281.28
    Each
       $1,281.28
    Supplier Number: 75706
  • 103302-164
    SEM Mount JEOL
    SEM Mount JEOL
    50 x 10 mm
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $157.88
    $157.88
    Each
       $157.88
    Supplier Number: 75707
  • 103306-182
    SEM Mount JEOL
    SEM Mount JEOL
    50 x 10 mm
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $1,392.25
    $1,392.25
    Each
       $1,392.25
    Supplier Number: 75708
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)