Order Entry
Canada
ContactUsLinkComponent
Tris-HCl Buffer, 1.5 M Sterile Solution, Teknova
Tris-HCl Buffer, 1.5 M Sterile Solution, Teknova
Catalog # CA101320-670
Supplier:  Teknova
CAS Number:  
Tris-HCl Buffer, 1.5 M Sterile Solution, Teknova
Catalog # CA101320-670
Supplier:  Teknova
Supplier Number:  T1588
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Tris-HCl Buffer 1.5 M
  • Size:
    1 L
  • Environmentally Preferable:
  • Storage temperature:
    Store at room temperature. Shipped at ambient temperature.
  • Cat. no.:
    CA101320-670
  • pH:
    8.8
  • Supplier No.:
    T1588

Specifications

About this item

1.5 M Tris (hydroxymethyl) Aminomethane solution adjusted to pH 8.8.