Order Entry
Canada
ContactUsLinkComponent
Human Recombinant MMP-9 (from E. coli)
Human Recombinant MMP-9 (from E. coli)
Supplier:  Anaspec Inc
Certificates
Human Recombinant MMP-9 (from E. coli)
Supplier:  Anaspec Inc
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –78 °C
  • Protein synonyms:
    MMP-9 Catalytic Domain|MMP-9|Matrix metalloproteinase 9
  • Protein/peptide name:
    MMP-9
  • UniProtKB:
    P14780
  • Purity:
    >95% as estimated by SDS-PAGE
  • Molecular weight:
    40 kDa

Specifications

Related Information

Frequently Bought Together