Order Entry
Canada
ContactUsLinkComponent
Human MMP-1 (from E. coli)
Human MMP-1 (from E. coli)
Catalog # 103010-276
Supplier:  Anaspec Inc
CAS Number:  
Human MMP-1 (from E. coli)
Catalog # 103010-276
Supplier:  Anaspec Inc
Supplier Number:  AS-72004
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µl
  • Storage conditions:
    –78 °C
  • Protein synonyms:
    MMP-1 Proenzyme|Matrix metalloproteinase 1|MMP-1 recombinant
  • UniProtKB:
    P03956
  • Protein/peptide name:
    MMP-1
  • Molecular weight:
    56kDa/52 kDa
  • Concentration:
    10 μg/mL
  • Cat. no.:
    103010-276
  • Supplier No.:
    AS-72004

Specifications

About this item

Matrix metalloproteinases (MMP’s) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-1 (collagenase-1) is involved in tumor development and metastasis and rheumatoid arthritis. It is proposed as a therapeutic target for these diseases. Native pro-MMP-1 is prepared from culture medium of human rheumatoid synovial fibroblasts. MMP-1 is secreted as pro-enzyme, which consists of a propeptide of 80 amino acids, a catalytic domain of 162 amino acids, a 16-residue linker region, and a hemopexin domain of 189 amino acids. The native pro-MMP-1 has a major Mr 52-kDa unglycosylated and a minor Mr 57-kDa glycosylated form. The proteolytic activation of the 57/52-kDa species will form 47/42-kDa active collagenase, and a 22-kDa C-terminal fragment
The apparent Mr on SDS-PAGE is approximately 56kDa/52 kDa. The pro-MMP-1 can be fully activated by incubating with 1 mM APMA at 37°C for 3 hr. Its activity can be measured by FRET peptides. 10-20 ng of enzyme is sufficient for FRET-based assay