Order Entry
Canada
ContactUsLinkComponent
Rat Recombinant Renin (from HEK293 Cells)
Rat Recombinant Renin (from HEK293 Cells)
Catalog # 103010-484
Supplier:  Anaspec Inc
CAS Number:  
Rat Recombinant Renin (from HEK293 Cells)
Catalog # 103010-484
Supplier:  Anaspec Inc
Supplier Number:  AS-72141
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    20 µg
  • Storage conditions:
    –78 °C
  • UniProtKB:
    P08424
  • Protein/peptide name:
    Renin
  • Molecular weight:
    38.5 kDa
  • Cat. no.:
    103010-484
  • Supplier No.:
    AS-72141

Specifications

About this item

Renin, a highly specific aspartyl protease, cleaves angiotensinogen, produced in the liver, to yield angiotensin I, which is further converted into angiotensin II by ACE (Angiotensin Converting Enzyme). Angiotensin II constricts blood vessels, leading to increased blood pressure. It also increases the secretion of ADH and aldosterone, and stimulates the hypothalamus to activate the thirst reflex. Since an overactive renin-angiotensin system leads to hypertension, renin is proposed as a therapeutic target for this disease.

Recombinant rat pro-renin was expressed in HEK cells. Purified enzyme was converted to the active renin by tryptic activation followed by removal of trypsin. The molecular mass of active rat renin is approximately 40 kDa. The activity of enzyme can be measured in FRET-based assays