Specifications
- Antibody type:Primary
- Antigen name:CCL18 Picoband™
- Antigen symbol:CCL18/PARC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:21 to 89
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human CCL18 recombinant protein (Position: A21-A89).
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-506
- Supplier No.:PB9023
Specifications
About this item
Polyclonal antibody for PARC/CCL18 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. PARC/CCL18 information: Molecular Weight: 9849 MW; Subcellular Localization: Secreted; Tissue Specificity: Expressed at high levels in lung, lymph nodes, placenta, bone marrow, dendritic cells present in germinal centers and T-cell areas of secondary lymphoid organs and macrophages derived from peripheral blood monocytes. Not expressed by peripheral blood monocytes and a monocyte-to-macrophage differentiation is a prerequisite for expression. Expressed in synovial fluids from patients with rheumatoid and septic arthritis and in ovarian carcinoma ascitic fluid.
Type: Primary
Antigen: CCL18/PARC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human