Anti-IKK Alpha Polyclonal Antibody
10209-320
- Antibody type:Primary
- Antigen name:IKK alpha Picoband™
- Antigen symbol:IKK Alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:411 to 745
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human IKK alpha recombinant protein (Position: V411-E745). Human IKK alpha shares 98% amino acid (aa) sequence identity with mouse IKK alpha.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-320
- Supplier No.:PB9110
Polyclonal antibody for IKK alpha/CHUK detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. IKK alpha/CHUK information: Molecular Weight: 84640 MW; Subcellular Localization: Cytoplasm . Nucleus . Shuttles between the cytoplasm and the nucleus; Tissue Specificity: Widely expressed.
Type: Primary
Antigen: IKK Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat