Anti-Cathepsin G Polyclonal Antibody
10209-276
- Antibody type:Primary
- Antigen name:Cathepsin G
- Antigen symbol:CTSG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CATG|CG
- Amino acid number:76 to 255
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human Cathepsin G recombinant protein (Position: A76-L255). Human Cathepsin G shares 66% amino acid (aa) sequence identity with mouse Cathepsin G.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-276
- Supplier No.:PB9049
Polyclonal antibody for CATHEPSIN G/CTSG detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. CATHEPSIN G/CTSG information: Molecular Weight: 28837 MW; Subcellular Localization: Cell surface .
Type: Primary
Antigen: Cathepsin G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat