Anti-Integrin Beta 4 Polyclonal Antibody
Catalog # 10206-222
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Integrin Beta 4
- Antigen symbol:ITGB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:28 to 266
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human Integrin beta 4 recombinant protein (Position: N28-A266). Human Integrin beta 4 shares 88% and 86% amino acid (aa) sequence identity with mouse and rat Integrin beta 4, respectively.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-222
- Supplier No.:PB9007
Specifications
About this item
Polyclonal antibody for CD104/ITGB4 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD104/ITGB4 information: Molecular Weight: 202167 MW; Subcellular Localization: Cell membrane; Single-pass type I membrane protein. Cell membrane; Lipid-anchor. Cell junction, hemidesmosome. Colocalizes with DST at the leading edge of migrating keratinocytes; Tissue Specificity: Integrin alpha-6/beta-4 is predominantly expressed by epithelia. Isoform beta-4D is also expressed in colon and placenta. Isoform beta-4E is also expressed in epidermis, lung, duodenum, heart, spleen and stomach.
Type: Primary
Antigen: Integrin Beta 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human