Magnetic Stir Bar, Giant, PTFE, Chemglass
Supplier: Chemglass
Certificates
About this item
These heavy duty PTFE stirring magnet designed for use with carboys up to 12 gallon.
- Capable of mixing volumes up to a 50 gallon drum
- Fits into a minimum neck diameter of 32 mm
Manufactured with FDA-grade and USP Class VI PTFE.
Customers Who Bought This Also Bought