K-kit Specimen Holder for Liquid TEM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
K-kits are enclosed specimen sample holders designed to facilitate convenient TEM observation of liquid samples, allowing nanoobjects, aggregates, and agglomerates (NOAAs) in liquid samples to be characterized.
- Can be used with most TEM holder brands
- Strong structural reliability under vacuum
- Sealing glue compatible to many solvents
Vacuum compatible sealing of liquids in electron-transmitting thickness, K-kits are micro reaction chambers for countless experiments in materials, chemical, and biological research.
Ordering information: Tools are optional available in a Tool Set or ordered individually. Glues are also available.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together