Autosampler Caps and Septa, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
PerkinElmer closures are manufactured to specific tolerances that are guaranteed to fit and perform with PerkinElmer and non-PerkinElmer instruments.
These septa have a very broad chemical resistance and can be used in many markets including: environmental, diagnostic packaging, pharmaceutical packaging, cosmetic and food packaging.
Frequently Bought Together