Order Entry
Canada
Orders LinkContactUsLinkComponent
Buffer, TE Solution (TRIS-EDTA Buffer) pH 8.0

Buffer, TE Solution (TRIS-EDTA Buffer) pH 8.0

TE, DEPC Treated, 1 L, 8.0
Catalog # CAMB-040
Supplier #:  MB-040
Buffer, TE Solution (TRIS-EDTA Buffer) pH 8.0
Catalog # CAMB-040
CAS Number:  
Buffer, TE Solution (TRIS-EDTA Buffer) pH 8.0

TE, DEPC Treated, 1 L, 8.0

CAS Number:  
Price
$203.16
Log in for availability
Each (1l)
Minimum order quantity is 1
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    TE, DEPC Treated
  • Size:
    1 L
  • pH:
    8.0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.