Order Entry
Canada
Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE, 50X, 1 L, 8.3
Catalog # CAMB-020
Supplier #:  MB-020
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # CAMB-020
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE, 50X, 1 L, 8.3

CAS Number:  
Price
$242.07
Log in for availability
Each (1l)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    TAE, 50X
  • Size:
    1 L
  • pH:
    8.3

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.