Pierce™ Protein L Plus Agarose, Thermo Scientific
CAPI20520
:
- Description:Protein L Plus Agarose
- Size:2 mL
- Binding capacity:10 – 20 mg Human IgG/mL Resin
- Cat. no.:CAPI20520
- Supplier No.:20520
Pierce™ Protein L Plus agarose is a high-capacity Protein L beaded agarose resin for use in a variety of antibody affinity purification methods.
- Immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Protein L is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non-specific binding and enabling multiple uses without decline in yield
- High capacity beads with a dense load of immobilized Protein L