Order Entry
Canada
ContactUsLinkComponent
EZ DNA Methylation™ Kits, Zymo Research
EZ DNA Methylation™ Kits, Zymo Research
  77001-564
 :  
undefined
EZ DNA Methylation™ Kits, Zymo Research
  77001-564
 :  D5004
 :  

 

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Methylated DNA
  • Sample type:
    DNA preps
  • Target molecule:
    DNA
  • Description:
    EZ-96 DNA Methylation™ Kit
  • Size:
    2 x 96 Preps
  • Description:
    EZ-96 DNA Methylation™ Kit
  • Cat. no.:
    77001-564
  • Format:
    96-well spin column plate

 

 

The EZ DNA Methylation™ Kits feature simplified procedures that streamline bisulfite treatment of DNA.

  • Streamlined, proven procedure for bisulfite conversion of DNA
  • Desulphonation and recovery of bisulfite-treated DNA with spin column, 96-well plate, and magnetic bead format
  • Recovered DNA is ideal for downstream analyses including PCR, endonuclease digestion, sequencing, microarrays, etc

The EZ DNA Methylation™ Kit features simplified procedure that streamline bisulfite treatment of DNA. This kit is the original bisulfite conversion kit from Zymo Research. The EZ DNA Methylation™ Kit is based on the three-step reaction that takes place between cytosine and sodium bisulfite where cytosine is converted into uracil. Innovative desulphonation technologies eliminate otherwise cumbersome precipitations. Designed to reduce template degradation, this kit minimizes DNA loss during treatment and cleanup, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc. These kits are recommended with Illumina’s GoldenGate® and Infinium® Assays.